CAS RN 50306-03-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-(-)-Clenbuterol, TRC
CAS: 50306-03-1 Molecular Formula: C12 H18 Cl2 N2 O Molecular Weight (g/mol): 277.19 Synonym: Benzenemethanol, 4-amino-3,5-dichloro-α-[[(1,1-dimethylethyl)amino]methyl]-, (αR)-,Benzenemethanol, 4-amino-3,5-dichloro-α-[[(1,1-dimethylethyl)amino]methyl]-, (R)-,(αR)-4-Amino-3,5-dichloro-α-[[(1,1-dimethylethyl)amino]methyl]benzenemethanol,(-)-Clenbuterol,(R)-Clenbuterol IUPAC Name: (1R)-1-(4-amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanol SMILES: CC(C)(C)NC[C@H](O)c1cc(Cl)c(N)c(Cl)c1