accessibility menu, dialog, popup

UserName

CAS RN 504-03-0

CH3(R)(S)H3CH2N

CAS RN 504-03-0

IUPAC Name: (2R,6S)-2,6-dimethylpiperidin-1-ium
Synonyms: (2R,6S)-2,6-dimethylpiperidin-1-ium
Molecular Weight (g/mol): 114.21
Molecular Formula: C7H16N
InChi Key: SDGKUVSVPIIUCF-KNVOCYPGSA-O
SMILES: C[C@H]1CCC[C@@H](C)[NH2+]1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results