accessibility menu, dialog, popup

UserName

CAS RN 5048-19-1

H2CN

CAS RN 5048-19-1

IUPAC Name: hex-5-enenitrile
Synonyms: hex-5-enenitrile
Molecular Weight (g/mol): 95.15
Molecular Formula: C6H9N
InChi Key: UNAQSRLBVVDYGP-UHFFFAOYSA-N
SMILES: C=CCCCC#N
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)