accessibility menu, dialog, popup

UserName

NClFFFOOH

CAS RN 505084-58-2

IUPAC Name: 2-chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid
Synonyms: 2-chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid
Molecular Weight (g/mol): 225.55
Molecular Formula: C7H3ClF3NO2
InChi Key: OIODRBHIESSYRO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=NC=C1C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results