accessibility menu, dialog, popup

UserName

CH3OONNBrSCH3

CAS RN 50593-91-4

IUPAC Name: methyl 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylate
Synonyms: methyl 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylate
Molecular Weight (g/mol): 263.11
Molecular Formula: C7H7BrN2O2S
InChi Key: MYZJIEWTRJTWCD-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(SC)=NC=C1Br
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)