accessibility menu, dialog, popup

UserName

CAS RN 506-51-4

H3COH

CAS RN 506-51-4

IUPAC Name: tetracosan-1-ol
Synonyms: 1-tetracosanol
Molecular Weight (g/mol): 354.66
Molecular Formula: C24H50O
InChi Key: TYWMIZZBOVGFOV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCO
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)