accessibility menu, dialog, popup

UserName

CAS RN 5072-26-4

CH3SHNONH2OHO

CAS RN 5072-26-4

IUPAC Name: 2-amino-4-[butyl(imino)oxo-λ⁶-sulfanyl]butanoic acid
Synonyms: buthionine sulfoximine buthionine sulphoximine butionine sulfoximine buthionine sulfoxamine buthione sulfoximine buthionine-S,R-sulfoximine sulfoximine, buthionine
Molecular Weight (g/mol): 222.3
Molecular Formula: C8H18N2O3S
InChi Key: KJQFBVYMGADDTQ-UHFFFAOYNA-N
SMILES: CCCCS(=N)(=O)CCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)