accessibility menu, dialog, popup

UserName

CAS RN 51-74-1

NH2NHNOHPOHOHOOHPOHOHO

CAS RN 51-74-1

IUPAC Name: 2-(1H-imidazol-5-yl)ethan-1-amine; bis(phosphoric acid)
Synonyms: histamine
Molecular Weight (g/mol): 307.14
Molecular Formula: C5H15N3O8P2
InChi Key: ZHIBQGJKHVBLJJ-UHFFFAOYSA-N
SMILES: OP(O)(O)=O.OP(O)(O)=O.NCCC1=CN=CN1
Certifications/Compliance 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results