accessibility menu, dialog, popup

UserName

ONO(E)

CAS RN 5153-67-3

IUPAC Name: [(1E)-2-nitroethenyl]benzene
Synonyms: β-nitrostyrene
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: PIAOLBVUVDXHHL-VOTSOKGWSA-N
SMILES: [O-][N+](=O)\C=C\C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)