accessibility menu, dialog, popup

UserName

H3COONHNH3C

CAS RN 51605-32-4

IUPAC Name: ethyl 5-methyl-1H-imidazole-4-carboxylate
Synonyms: ethyl 5-methyl-1H-imidazole-4-carboxylate
Molecular Weight (g/mol): 154.17
Molecular Formula: C7H10N2O2
InChi Key: VLDUBDZWWNLZCU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)NC=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results