accessibility menu, dialog, popup

UserName

CAS RN 51770-71-9

IUPAC Name: 2,4-dimethoxybenzene-1-sulfonamide
Synonyms: 2,4-dimethoxybenzenesulfonamide
Molecular Weight (g/mol): 217.24
Molecular Formula: C8H11NO4S
InChi Key: MGHCDRVTMABICG-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results