CAS RN 518357-43-2
IUPAC Name:
1-(4-chloro-2,6-difluorophenyl)methanamine
Synonyms:
1-(4-chloro-2,6-difluorophenyl)methanamine
Molecular Weight (g/mol):
177.58
Molecular Formula:
C7H6ClF2N
InChi Key:
YBILGZZEXORCAH-UHFFFAOYSA-N
SMILES:
NCC1=C(F)C=C(Cl)C=C1F
1
–
1
of
1
results
1
4-Chloro-2,6-difluorobenzylamine, 97%, Thermo Scientific™
CAS: 518357-43-2 Molecular Formula: C7H6ClF2N Molecular Weight (g/mol): 177.579 MDL Number: MFCD06660350 InChI Key: YBILGZZEXORCAH-UHFFFAOYSA-N Synonym: 4-chloro-2,6-difluorobenzylamine,4-chloro-2,6-difluorophenyl methanamine,benzenemethanamine, 4-chloro-2,6-difluoro,2,6-difluoro-4-chlorobenzylamine,4-chloro-2,6-difluorophenyl methylamine,4-chloro-2,6-difluoro-phenyl methanamine,1-4-chloro-2,6-difluorophenyl methanamine PubChem CID: 17750831 IUPAC Name: (4-chloro-2,6-difluorophenyl)methanamine SMILES: C1=C(C=C(C(=C1F)CN)F)Cl