accessibility menu, dialog, popup

UserName

NH2HN

CAS RN 5192-03-0

IUPAC Name: 1H-indol-5-amine
Synonyms: 5-aminoindole
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: ZCBIFHNDZBSCEP-UHFFFAOYSA-N
SMILES: NC1=CC=C2NC=CC2=C1
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)