accessibility menu, dialog, popup

UserName

H3CH3CSH(S)NH2OHO

CAS RN 52-67-5

IUPAC Name: (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
Synonyms: penicillamine cupripen (D)-penicillamine artamine penicillamin (S)-penicillamine pendramine penicillaminate, copper dimethylcysteine D-penicilamine
Molecular Weight (g/mol): 149.21
Molecular Formula: C5H11NO2S
InChi Key: VVNCNSJFMMFHPL-VKHMYHEASA-N
SMILES: CC(C)(S)[C@@H](N)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)