CAS RN 522-27-0
IUPAC Name:
N-[(1E)-1,2-bis(furan-2-yl)-2-nitrosoethenyl]hydroxylamine
Synonyms:
N-[(1E)-1,2-bis(furan-2-yl)-2-nitrosoethenyl]hydroxylamine
Molecular Weight (g/mol):
220.18
Molecular Formula:
C10H8N2O4
InChi Key:
BRKZOASOLMUAQJ-MDZDMXLPSA-N
SMILES:
ON\C(=C(\N=O)C1=CC=CO1)C1=CC=CO1
1
–
1
of
1
results
1
2,2'-Furil dioxime, mixture of isomers, 97%
CAS: 522-27-0 Molecular Formula: C10H8N2O4 Molecular Weight (g/mol): 220.18 MDL Number: MFCD00075315 InChI Key: RBOZTFPIXJBLPK-DSOJMZEYSA-N Synonym: alpha-furil dioxime,furildioxime,alpha-furyldioxime,di-2-furylglyoxime,bis 2-furyl glyoxime,di-2-furylethanedione dioxime,ethanedione, di-2-furanyl-, dioxime,glyoxime, di-2-furyl,furil, dioxime,1z,2z-1,2-di furan-2-yl ethane-1,2-dione dioxime PubChem CID: 5359431 SMILES: O\N=C(\C(=N/O)\C1=CC=CO1)/C1=CC=CO1