CAS RN 52250-50-7
CAS RN 52250-50-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Phenyl-3,4-dihydro-isoquinoline, TRC
CAS: 52250-50-7 Molecular Formula: C15H13N Molecular Weight (g/mol): 207.27 Synonym: 3,4-Dihydro-1-phenylisoquinoline,1-Phenyl-3,4-dihydroisoquinoline IUPAC Name: 1-phenyl-3,4-dihydroisoquinoline SMILES: C1Cc2ccccc2C(=N1)c3ccccc3