CAS RN 52320-67-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methylbutyrylglycine, TRC
CAS: 52320-67-9 Molecular Formula: C7 H13 N O3 Molecular Weight (g/mol): 159.18 Synonym: Glycine, N-(2-methyl-1-oxobutyl)-,N-(2-Methyl-1-oxobutyl)glycine,2-Methylbutyrylglycine,N-(2-Methylbutyryl)glycine,N-sec-Valerylglycine,α-Methylbutyrylglycine,2-(2-Methylbutanoylamino)acetic acid,N-(2-Methylbutanoyl)glycine IUPAC Name: 2-(2-methylbutanoylamino)acetic acid SMILES: CCC(C)C(=O)NCC(=O)O