CAS RN 52373-72-5
CAS RN 52373-72-5
IUPAC Name:
methyl 2,2-dimethyl-1,3-dioxolane-4-carboxylate
Synonyms:
methyl 2,2-dimethyl-1,3-dioxolane-4-carboxylate
Molecular Weight (g/mol):
160.17
Molecular Formula:
C7H12O4
InChi Key:
DOWWCCDWPKGNGX-UHFFFAOYNA-N
SMILES:
COC(=O)C1COC(C)(C)O1
1
–
3
of
3
results
1
(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.