accessibility menu, dialog, popup

UserName

CAS RN 52497-36-6

CH3H3C(R)NH(S)(S)OHOOHO

CAS RN 52497-36-6

IUPAC Name: (2S,3S,4R)-3-(carboxymethyl)-4-(propan-2-yl)pyrrolidine-2-carboxylic acid
Synonyms: dihydrokainate dihydrokainic acid
Molecular Weight (g/mol): 215.25
Molecular Formula: C10H17NO4
InChi Key: JQPDCKOQOOQUSC-OOZYFLPDSA-N
SMILES: CC(C)[C@H]1CN[C@@H]([C@H]1CC(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results