accessibility menu, dialog, popup

UserName

CAS RN 5271-27-2

CH3NHNH2(R)

CAS RN 5271-27-2

IUPAC Name: (2R)-4-methyl-2-phenylpiperazine-1,4-diium
Synonyms: (2R)-4-methyl-2-phenylpiperazine-1,4-diium
Molecular Weight (g/mol): 178.28
Molecular Formula: C11H18N2
InChi Key: IRMBVBDXXYXPEW-NSHDSACASA-P
SMILES: C[NH+]1CC[NH2+][C@@H](C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)