CAS RN 52806-53-8
IUPAC Name:
2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
Synonyms:
hydroxyflutamide
Molecular Weight (g/mol):
292.21
Molecular Formula:
C11H11F3N2O4
InChi Key:
YPQLFJODEKMJEF-UHFFFAOYSA-N
SMILES:
CC(C)(O)C(=O)NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
1
–
3
of
3
results
1
2-Hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide (2-Hydroxyflutamide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Hydroxy Flutamide, TRC
CAS: 52806-53-8 Molecular Formula: C11 H11 F3 N2 O4 Molecular Weight (g/mol): 292.21 Synonym: 2-Hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide,2-Hydroxyflutamide IUPAC Name: 2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide SMILES: CC(C)(O)C(=O)Nc1ccc(c(c1)C(F)(F)F)[N+](=O)[O-]