accessibility menu, dialog, popup

UserName

CH3O

CAS RN 529-20-4

IUPAC Name: 2-methylbenzaldehyde
Synonyms: 2-methylbenzaldehyde O-tolualdehyde o-methylbenazldehyde toluic aldehyde
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: BTFQKIATRPGRBS-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (3)