accessibility menu, dialog, popup

UserName

CAS RN 53130-43-1

FFFClOFFF

CAS RN 53130-43-1

IUPAC Name: 2,4-bis(trifluoromethyl)benzoyl chloride
Synonyms: 2,4-bis(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 276.56
Molecular Formula: C9H3ClF6O
InChi Key: TZXHEDOCQVTEPT-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C(Cl)=O)C(=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results