CAS RN 5324-47-0
CAS RN 5324-47-0
IUPAC Name:
sodium ethanesulfonate
Synonyms:
sodium esylate
Molecular Weight (g/mol):
132.11
Molecular Formula:
C2H5NaO3S
InChi Key:
KQFAFFYKLIBKDE-UHFFFAOYSA-M
SMILES:
[Na+].CCS([O-])(=O)=O
1
–
1
of
1
results
1
Ethanesulfonic acid sodium salt, 98%, may cont. ca 2% water
CAS: 5324-47-0 Molecular Formula: C2H5NaO3S Molecular Weight (g/mol): 132.109 MDL Number: MFCD00066498 InChI Key: KQFAFFYKLIBKDE-UHFFFAOYSA-M Synonym: sodium ethanesulfonate,sodium ethanesulphonate,ethanesulfonic acid sodium salt,ethanesulfonic acid, sodium salt,ethane sulfonic acid sodium salt,sodiumethanesulfonate,sodium ethylsulfonate,sodium esylate,sodium ethane sulfonate,ethylsulfonic acid sodium salt PubChem CID: 4192170 IUPAC Name: sodium;ethanesulfonate SMILES: CCS(=O)(=O)[O-].[Na+]