accessibility menu, dialog, popup

UserName

CAS RN 53293-00-8

OOCH

CAS RN 53293-00-8

IUPAC Name: hex-5-ynoate
Synonyms: hex-5-ynoate
Molecular Weight (g/mol): 111.12
Molecular Formula: C6H7O2
InChi Key: VPFMEXRVUOPYRG-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCCC#C
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)