accessibility menu, dialog, popup

UserName

CAS RN 5332-06-9

BrN

CAS RN 5332-06-9

IUPAC Name: 4-bromobutanenitrile
Synonyms: butanenitrile, 4-bromo-
Molecular Weight (g/mol): 148
Molecular Formula: C4H6BrN
InChi Key: CQPGDDAKTTWVDD-UHFFFAOYSA-N
SMILES: BrCCCC#N
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)