accessibility menu, dialog, popup

UserName

NH2SNHCl

CAS RN 5344-82-1

IUPAC Name: (2-chlorophenyl)thiourea
Synonyms: (2-chlorophenyl)thiourea
Molecular Weight (g/mol): 186.66
Molecular Formula: C7H7ClN2S
InChi Key: YZUKKTCDYSIWKJ-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results