accessibility menu, dialog, popup

UserName

H3COOCH3Cl

CAS RN 535-13-7

IUPAC Name: ethyl 2-chloropropanoate
Synonyms: ethyl α-chloropropionate
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: JEAVBVKAYUCPAQ-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)Cl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)