CAS RN 53636-19-4
IUPAC Name:
1-chloro-1-oxopropan-2-yl acetate
Synonyms:
1-chloro-1-oxopropan-2-yl acetate
Molecular Weight (g/mol):
150.56
Molecular Formula:
C5H7ClO3
InChi Key:
ALHZEIINTQJLOT-UHFFFAOYNA-N
SMILES:
CC(OC(C)=O)C(Cl)=O
1
–
3
of
3
results
1
(R)-(+)-2-Acetoxypropionyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-(+)-2-Acetoxypropionyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R)-(+)-2-Acetoxypropionyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.