accessibility menu, dialog, popup

UserName

H3COH3CNOO

CAS RN 5367-32-8

IUPAC Name: 4-methoxy-2-methyl-1-nitrobenzene
Synonyms: 3-methyl-4-nitroanisole
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: RTZOGYCMIMOVHU-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(C)=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results