accessibility menu, dialog, popup

UserName

FOFBr

CAS RN 537013-51-7

IUPAC Name: 4-bromo-2,6-difluorobenzaldehyde
Synonyms: 4-bromo-2,6-difluorobenzaldehyde
Molecular Weight (g/mol): 221
Molecular Formula: C7H3BrF2O
InChi Key: CZGVAISJIQNQEJ-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(F)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results