accessibility menu, dialog, popup

UserName

CH3NOH3CCH3(E/Z)(E/Z)

CAS RN 53704-25-9

IUPAC Name: 2-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 2-[2-(1,3,3-trimethylindol-2-ylidene)ethylidene]-3,4-dihydronaphthalen-1-one
Molecular Weight (g/mol): 329.44
Molecular Formula: C23H23NO
InChi Key: DTPLBBMEWNNVFO-UHFFFAOYSA-N
SMILES: CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCC2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results