accessibility menu, dialog, popup

UserName

CAS RN 538-39-6

CH3CH3

CAS RN 538-39-6

IUPAC Name: 1-methyl-4-[2-(4-methylphenyl)ethyl]benzene
Synonyms: 1-methyl-4-[2-(4-methylphenyl)ethyl]benzene
Molecular Weight (g/mol): 210.32
Molecular Formula: C16H18
InChi Key: XCCQFUHBIRHLQT-UHFFFAOYSA-N
SMILES: CC1=CC=C(CCC2=CC=C(C)C=C2)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)