accessibility menu, dialog, popup

UserName

CAS RN 53871-27-5

IUPAC Name: 1-(3-fluorophenyl)-1H-pyrrole
Synonyms: 1-(3-fluorophenyl)pyrrole
Molecular Weight (g/mol): 161.18
Molecular Formula: C10H8FN
InChi Key: MMILNBLSOHVLCJ-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1)N1C=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results