CAS RN 539-15-1
CAS RN 539-15-1
IUPAC Name:
4-[2-(dimethylamino)ethyl]phenol; sulfuric acid
Synonyms:
hordenine
Molecular Weight (g/mol):
263.31
Molecular Formula:
C10H17NO5S
InChi Key:
OIIQUBZPQJNHQK-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.CN(C)CCC1=CC=C(O)C=C1
1
–
1
of
1
results
1
Hordenine, TRC
CAS: 539-15-1 Molecular Formula: C10 H15 N O Molecular Weight (g/mol): 165.23 Synonym: Eremursine (7CI),Hordenine (6CI),Phenol, p-[2-(dimethylamino)ethyl]- (8CI),4-[2-(Dimethylamino)ethyl]phenol,Anhalin,Anhaline,Cactine,Hordenin,N,N-Dimethyl-4-hydroxy-β-phenethylamine,N,N-Dimethyl-p-hydroxyphenethylamine,N,N-Dimethyltyramine,Ordenina,Peyocactine,p-Hydroxy-N,N-dimethylphenethylamine,p-[2-(Dimethylamino)ethyl]phenol IUPAC Name: 4-[2-(dimethylamino)ethyl]phenol SMILES: CN(C)CCc1ccc(O)cc1