CAS RN 5391-39-9
IUPAC Name:
1-acetylimidazolidin-2-one
Synonyms:
1-acetylimidazolidin-2-one
Molecular Weight (g/mol):
128.13
Molecular Formula:
C5H8N2O2
InChi Key:
JJWACYUTERPMBM-UHFFFAOYSA-N
SMILES:
CC(=O)N1CCNC1=O
1
–
2
of
2
results
1
1-Acetyl-2-imidazolidinone, TRC
CAS: 5391-39-9 Molecular Formula: C5 H8 N2 O2 Molecular Weight (g/mol): 128.13 Synonym: 1-Acetylimidazolidin-2-one,Clonidine Hydrochloride Imp. A (EP),2-Imidazolidinone, 1-acetyl-,1-Acetyl-2-imidazolidinone,N-Acetyl-2-imidazolidinone,N-Acetylimidazolidinone,N-Acetylethylene urea IUPAC Name: 1-acetylimidazolidin-2-one SMILES: CC(=O)N1CCNC1=O
2
1-Acetylimidazolidin-2-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.