CAS RN 53990-33-3
IUPAC Name:
2-{[(benzyloxy)carbonyl]amino}-2-phenylacetic acid
Synonyms:
{[(benzyloxy)carbonyl]amino}(phenyl)acetic acid
Molecular Weight (g/mol):
285.3
Molecular Formula:
C16H15NO4
InChi Key:
RLDJWBVOZVJJOS-UHFFFAOYNA-N
SMILES:
OC(=O)C(NC(=O)OCC1=CC=CC=C1)C1=CC=CC=C1
1
–
3
of
3
results
1
Z-L-Phenylglycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Z-L-Phenylglycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Z-L-Phenylglycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.