accessibility menu, dialog, popup

UserName

CAS RN 53994-73-3

H2N(R)OHN(R)(R)SNOOHOCl

CAS RN 53994-73-3

IUPAC Name: (6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: cefaclor cefaclorum
Molecular Weight (g/mol): 367.8
Molecular Formula: C15H14ClN3O4S
InChi Key: QYIYFLOTGYLRGG-GPCCPHFNSA-N
SMILES: N[C@@H](C(=O)N[C@H]1[C@H]2SCC(Cl)=C(N2C1=O)C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Percent Purity 
  • (2)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)