accessibility menu, dialog, popup

UserName

CAS RN 5402-55-1

OHS

CAS RN 5402-55-1

IUPAC Name: 2-(thiophen-2-yl)ethan-1-ol
Synonyms: 2-(thiophen-2-yl)ethanol
Molecular Weight (g/mol): 128.19
Molecular Formula: C6H8OS
InChi Key: VMJOFTHFJMLIKL-UHFFFAOYSA-N
SMILES: OCCC1=CC=CS1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)