accessibility menu, dialog, popup

UserName

CAS RN 541-73-1

ClCl

CAS RN 541-73-1

IUPAC Name: 1,3-dichlorobenzene
Synonyms: 3-dichlorobenzene m-phenylene dichloride metadichlorobenzene m-dcb m-phenylenedichloride m-dichlorobenzene (substance)
Molecular Weight (g/mol): 147
Molecular Formula: C6H4Cl2
InChi Key: ZPQOPVIELGIULI-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (5)
Boiling Point 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (5)
  • (3)
  • (7)
  • (5)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)