CAS RN 5438-07-3
CAS RN 5438-07-3
IUPAC Name:
2-amino-2-phenylbutanoic acid
Synonyms:
2-amino-2-phenylbutanoic acid
Molecular Weight (g/mol):
179.22
Molecular Formula:
C10H13NO2
InChi Key:
UBXUDSPYIGPGGP-UHFFFAOYNA-N
SMILES:
CCC(N)(C(O)=O)C1=CC=CC=C1
1
–
3
of
3
results
1
2-Amino-2-phenylbutyric Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Amino-2-phenylbutyric Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Amino-2-phenylbutyric Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.