accessibility menu, dialog, popup

UserName

CAS RN 54512-75-3

IUPAC Name: 1-bromo-5-chloropentane
Synonyms: 1-bromo-5-chloropentane
Molecular Weight (g/mol): 185.49
Molecular Formula: C5H10BrCl
InChi Key: PHHNNDKXQVKJEP-UHFFFAOYSA-N
SMILES: ClCCCCCBr
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)