CAS RN 5456-63-3
CAS RN 5456-63-3
IUPAC Name:
(1R,2R)-2-aminocyclohexan-1-ol hydrochloridyl
Synonyms:
(1R,2R)-2-aminocyclohexan-1-ol hydrochloridyl
Molecular Weight (g/mol):
150.63
Molecular Formula:
C6H13ClNO
InChi Key:
SEBSRZIHRZRNAH-KGZKBUQUSA-N
SMILES:
[Cl].N[C@@H]1CCCC[C@H]1O
1
–
2
of
2
results
1
trans-2-Aminocyclohexanol hydrochloride, 99%
CAS: 5456-63-3 Molecular Formula: C6H13ClNO Molecular Weight (g/mol): 150.63 MDL Number: MFCD00001490,MFCD09259962,MFCD09259962,MFCD09259963,MFCD11618002,MFCD11618003,MFCD09259963 InChI Key: SEBSRZIHRZRNAH-KGZKBUQUSA-N PubChem CID: 12228412 SMILES: [Cl].N[C@@H]1CCCC[C@H]1O
2
trans-2-Aminocyclohexanol hydrochloride, 97+%
CAS: 5456-63-3 Molecular Formula: C6H13ClNO Molecular Weight (g/mol): 150.63 MDL Number: MFCD00001490,MFCD09259962,MFCD09259962,MFCD09259963,MFCD11618002,MFCD11618003,MFCD09259963 InChI Key: SEBSRZIHRZRNAH-KGZKBUQUSA-N PubChem CID: 12228412 IUPAC Name: (1R,2R)-2-aminocyclohexan-1-ol hydrochloridyl SMILES: [Cl].N[C@@H]1CCCC[C@H]1O