accessibility menu, dialog, popup

UserName

CAS RN 54745-74-3

NHOHCl

CAS RN 54745-74-3

IUPAC Name: 8-oxa-3-azabicyclo[3.2.1]octane hydrochloride
Synonyms: 8-oxa-3-azabicyclo[3.2.1]octane hydrochloride
Molecular Weight (g/mol): 149.62
Molecular Formula: C6H12ClNO
InChi Key: XADOTNAXKKFKDY-UHFFFAOYNA-N
SMILES: Cl.C1CC2CNCC1O2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results