accessibility menu, dialog, popup

UserName

CAS RN 550-82-3

OHOONO

CAS RN 550-82-3

IUPAC Name: 7-hydroxy-10-oxo-10H-10λ⁵-phenoxazin-10-ylium-3-olate
Synonyms: resazurin
Molecular Weight (g/mol): 229.19
Molecular Formula: C12H7NO4
InChi Key: AUCGXRDZPQKALD-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC3=CC([O-])=CC=C3[N+]2=O)=C1
Color 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)