CAS RN 55142-16-0
IUPAC Name:
methyl 2-(benzyloxy)benzoate
Synonyms:
methyl 2-(benzyloxy)benzoate
Molecular Weight (g/mol):
242.27
Molecular Formula:
C15H14O3
InChi Key:
FULHVBYAYSGTKX-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=CC=C1OCC1=CC=CC=C1
1
–
1
of
1
results
1
Methyl 2-benzyloxybenzoate, 98%, Thermo Scientific™
CAS: 55142-16-0 Molecular Formula: C15H14O3 Molecular Weight (g/mol): 242.27 MDL Number: MFCD01099045 InChI Key: FULHVBYAYSGTKX-UHFFFAOYSA-N Synonym: methyl 2-benzyloxybenzoate,methyl 2-benzyloxy benzoate,2-benzyloxybenzoic acid methyl ester,methyl 2-phenylmethoxy benzoate,methyl 2-benzyloxy-benzoate,acmc-1b03z,methyl salicylate monobenzyl ether,2-benzyloxy benzoic acid methyl ester,2-phenylmethoxybenzoic acid methyl ester,benzoic acid,2-phenylmethoxy-, methyl ester PubChem CID: 251331 IUPAC Name: methyl 2-phenylmethoxybenzoate SMILES: COC(=O)C1=CC=CC=C1OCC1=CC=CC=C1