accessibility menu, dialog, popup

UserName

CAS RN 5522-66-7

H3CHNNNNHH3COOH3COOCH3CH3CH2CH3H2C

CAS RN 5522-66-7

IUPAC Name: methyl 3-[9,14-diethenyl-20-(3-methoxy-3-oxopropyl)-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(21),2,4,6(24),7,9,11,13,15,17,19-undecaen-4-yl]propanoate
Synonyms: methyl 3-[9,14-diethenyl-20-(3-methoxy-3-oxopropyl)-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(21),2,4,6(24),7,9,11,13,15,17,19-undecaen-4-yl]propanoate
Molecular Weight (g/mol): 590.72
Molecular Formula: C36H38N4O4
InChi Key: LQBPATQBTSBIIH-UHFFFAOYSA-N
SMILES: COC(=O)CCC1=C(C)C2=NC1=CC1=NC(=CC3=C(C)C(C=C)=C(N3)C=C3NC(=C2)C(C=C)=C3C)C(C)=C1CCC(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)