accessibility menu, dialog, popup

UserName

OHNONOO(E)

CAS RN 555-15-7

IUPAC Name: (E)-N-[(5-nitrofuran-2-yl)methylidene]hydroxylamine
Synonyms: nifuroxime
Molecular Weight (g/mol): 156.1
Molecular Formula: C5H4N2O4
InChi Key: PTBKFATYSVLSSD-ZZXKWVIFSA-N
SMILES: O\N=C\C1=CC=C(O1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results