accessibility menu, dialog, popup

UserName

CAS RN 5619-07-8

ONH2OCH3HCl

CAS RN 5619-07-8

IUPAC Name: methyl 2-amino-3-phenylpropanoate hydrochloride
Synonyms: methyl 2-amino-3-phenylpropanoate hydrochloride
Molecular Weight (g/mol): 215.68
Molecular Formula: C10H14ClNO2
InChi Key: SWVMLNPDTIFDDY-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)