accessibility menu, dialog, popup

UserName

CAS RN 56309-94-5

OOO

CAS RN 56309-94-5

IUPAC Name: 4-{1,4-dioxaspiro[4.5]decan-8-yl}cyclohexan-1-one
Synonyms: 4-{1,4-dioxaspiro[4.5]decan-8-yl}cyclohexan-1-one
Molecular Weight (g/mol): 238.33
Molecular Formula: C14H22O3
InChi Key: ZNWLFTSPNBLXGL-UHFFFAOYSA-N
SMILES: O=C1CCC(CC1)C1CCC2(CC1)OCCO2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results